Learn More
Abnova™ Oxaloacetate Assay Kit
Used for quantifying OAA and functional study
Brand: Abnova™ KA3793.1Kit
Additional Details : Weight : 0.00010kg
Specifications
Colorimetric: 7 to 400μM, Fluorometric: 1 to 40μM | |
Colorimetric, Fluorometric | |
1 Kit | |
Oxaloacetate Assay Kit |
Oxaloacetate Assay Kit is a quantitative colorimetric/fluorimetric determination of oxaloacetate at OD570, FL530/585 nm. | |
Functional Study | |
Store the kit at -20°C. |