Learn More
Abnova™ Oxaloacetate Assay Kit
Used for quantifying OAA and functional study
Brand:  Abnova™ KA3793.1Kit
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16059049
  • 4298.00 SEK / 1 set
Estimated Shipment
03-02-2021
Specifications
Colorimetric: 7 to 400μM, Fluorometric: 1 to 40μM
Colorimetric, Fluorometric
1 Kit
Oxaloacetate Assay Kit
Oxaloacetate Assay Kit is a quantitative colorimetric/fluorimetric determination of oxaloacetate at OD570, FL530/585 nm.
Functional Study
Store the kit at -20°C.