missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPBP (Human) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human PPBP
POA
Specifications
Detection Method | Colorimetric |
---|---|
Sample Volume | 100 μL |
Content And Storage | Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |
Specifications
Colorimetric | |
100 μL | |
Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |