missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPBP (Human) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human PPBP
Brand: Abnova™ KA1753.1Kit
Additional Details : Weight : 0.02000kg
Specifications
Biotin | |
PPBP (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human PPBP. | |
Colorimetric | |
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solut | |
96 Assays | |
100Glc muL | |
100μL |
Human | |
8.5pg/mL | |
Quantification | |
PPBP (Human) ELISA Kit | |
Cell Culture Supernatant, Plasma, Serum, Urine | |
Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |