missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Progesterone ELISA Kit

Brand:  Abnova™ KA0299.1Kit

Additional Details : Weight : 0.02000kg


 View more versions of this product

Product Code. 16066015

  • 3848.00 SEK / 1 set


Estimated Shipment
09-02-2021
Add to basket

Includes: Goat anti-Mouse IgG Plate, 96 well
Progesterone Alkaline
Phosphatase Conjugate, 6mL
Progesterone Monoclonal Antibody, 6mL
Assay Buffer, 30mL
Wash Buffer Concentrate, 30mL
Progesterone Standard, 0.5mL
Steroid Displacement
Reagent, 1mL
pNpp Substrate, 20mL
Stop Solution, 5mL
Plate Sealer, 1 slice

Description

Description

Progesterone ELISA kit is a competitive immunoassay for quantitative determination of Progesterone in biological fluids
Specifications

Specifications

15.62 to 500pg/mL
Progesterone ELISA Kit is a competitive immunoassay for the quantitative determination of Progesterone in biological fluids.
Colorimetric
96 wells
Progesterone ELISA Kit
1 Kit
Biological fluid
8.57pg/mL
96 Assays
AP
8.57pg/mL
Quantification
Microwells (96 wells), EIA conjugate (5mL), EIA antibody (5mL), Assay buffer (27mL), Wash buffer concentrate (27mL), Standard (0.5mL), pNpp substrate (20mL), Stop solution (5mL)
ELISA Kit
RUO
100μL
Store the kit at 4°C.
100μL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Progesterone ELISA Kit

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide, sodiumphosphate tribasic

SDS
Documents