missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Prostaglandin F2 alpha ELISA Kit
Brand: Abnova™ KA0327.1Kit
Additional Details : Weight : 0.02000kg
Includes: Donkey anti-Sheep IgG Microtiter Plate, 96 Wells
PGF2α HS ELISA Conjugate, 2.5mL
PGF2α HS ELISA Antibody, 2.5mL
Assay Buffer, 30mL
Wash Buffer Concentrate, 27mL
Prostaglandin F2α Standard, 0.5mL
pNpp Substrate, 20mL
Stop Solution, 5mL
Plate Sealer, 1 slide
Description
The Prostaglandin F2 alpha ELISA Kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2α in milk, culture supernates, saliva, urine, serum and plasmaSpecifications
1.95 to 2000pg/mL | |
PGF2-alpha High Sensitivity ELISA Kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2-alpha in milk, culture supernates, saliva, urine, serum and plasma. | |
Colorimetric | |
96 wells | |
Prostaglandin F2 alpha ELISA Kit | |
1 Kit | |
Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine | |
0.98pg/mL | |
96 Assays |
AP | |
0.98pg/mL | |
Quantification | |
Microwells (96 wells), EIA Conjugate (5mL), EIA Antibody (5mL), Assay Buffer (30mL), Wash Buffer Concentrate (30mL), Standard (0.5mL), pNpp Substrate (20mL), Stop Solution: (5mL) | |
ELISA Kit | |
For research use only (RUO) | |
100μL | |
Store the kit at 4°C. | |
100μL |