missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RBP ELISA Kit

Brand:  Abnova™ KA3386.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16057965

  • 5943.00 SEK / 1 set


Estimated Shipment
11-02-2021
Add to basket

Includes: Coated Clear 96 Well Plate: A clear plastic microplate with break-apart strips coated with goat anti-rabbit IgG, 1 plate
RBP Standard: A stock solution of native human RBP at 20µg/mL, 60µL
RBP Antibody: A polyclonal antibody specific for RBP, 3mL
RBP-Peroxidase Conjugate: A RBP-peroxidase conjugate, 3mL
Assay Buffer, 28mL
Wash Buffer Concentrate: A 20X concentrate, 30mL
TMB Substrate, 11mL
Stop Solution: A 1N hydrochloric acid solution. Caustic, 11mL
Plate Sealer, 1 unit

Description

Description

The RBP ELISA Kit is designed to quantitatively measure RBP present in urine samples
Specifications

Specifications

0 to 1000ng/mL
Dog, Human, Monkey, Rat
4.09ng/mL
Quantification
Coated Clear 96 Well Plate: A clear plastic microplate with break-apart strips coated with goat anti-rabbit IgG (1plate), RBP Standard: A stock solution of native human RBP at 20μg/mL (60μL), RBP Antibody: A polyclonal antibody specific for RBP (3mL), RBP-Peroxidase Conjugate: A RBP-peroxidase conjugate (5mL), Assay Buffer (28mL ), Wash Buffer Concentrate: A 20X concentrate that should be diluted with deionized or distilled water (30mL), TMB Substrate (11mL), Stop Solution: A 1N hydrochloric acid solution. Caustic (11mL), Plate Sealer 1 each
For research use only (RUO)
50μL
Store the kit at 4°C.
50μL
HRP
RBP ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of RBP in human, rat, dog and rhesus monkey urine.
Colorimetric
96 wells
RBP ELISA Kit
Urine
2.90ng/mL
96 Assays
Videos
SDS
Documents