missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RBP ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | HRP (Horseradish Peroxidase) |
Product Type | RBP ELISA Kit |
Regulatory Status | For research use only (RUO) |
Content And Storage | Store the kit at 4°C |
Description
The RBP ELISA Kits are designed to quantitatively measure RBP present in serum, plasma, and urine samples.Specifications
Colorimetric | |
RBP ELISA Kit | |
Store the kit at 4°C |
HRP (Horseradish Peroxidase) | |
For research use only (RUO) |