missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RBP4 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human RBP4
Brand: Abnova™ KA0989.1Kit
Additional Details : Weight : 0.02000kg
Specifications
312 to 20000pg/mL | |
Human | |
10pg/mL | |
Quantification | |
RBP4 (Human) ELISA Kit | |
1 Kit | |
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate | |
96 Assays |
Biotin | |
RBP4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human RBP4. | |
Colorimetric | |
96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Human RBP4 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human RBP4 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin -Biotin | |
RBP4 (Human) ELISA kit | |
For research use only (RUO) | |
Store the kit at-20°C. | |
0.1mL |