missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SCGB1A1 (Human) ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Human |
Product Type | ELISA Kit |
Regulatory Status | For research use only (RUO) |
Sample Volume | 100 μL |
Description
SCGB1A1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SCGB1A1.Specifications
Colorimetric | |
ELISA Kit | |
100 μL |
Human | |
For research use only (RUO) |