missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SCGB1A1 (Human) ELISA Kit
Brand: Abnova™ KA0020.1Kit
Additional Details : Weight : 0.02000kg
Includes: Antibody Coated Microtiter Strips, 96 wells
Biotin Labelled Antibody, 13mL
Streptavidin-HRP Conjugate, 13mL
Master Standard, 2 vials
Quality Control HIGH, 2 vials
Quality Control LOW, 2 vials
Dilution Buffer, 20mL
Wash Solution Conc. (10x), 100mL
Substrate Solution, 13mL
Stop Solution, 13mL
Description
- The kit measures Clara cell protein in serum and plasma (EDTA, citrate, heparin)
- Quality Controls are human serum based
- Standard is recombinant protein (E. coli) based
- Components of the kit are provided ready to use, concentrated or lyophilized
Specifications
1.57 to 50ng/mL | |
Human | |
46pg/mL | |
Quantification | |
Antibody Coated Microtiter Strips 96 wells, Biotin Labelled Antibody (13mL), Streptavidin-HRP Conjugate (13mL), Master Standard (1 vial), Quality Control HIGH (2 vials), Quality Control LOW (2 vials), Dilution Buffer (20mL), Wash Solution Conc. (10x) (100mL), Substrate Solution (13mL), Stop Solution (13mL) | |
ELISA Kit | |
For research use only (RUO) | |
100μL | |
Store the kit at 4°C. | |
100μL |
HRP | |
SCGB1A1 (Human) ELISA Kit is a biotin labelled antibody based sandwich enzyme immunoassay for the quantitative measurement of human club cell protein. | |
Colorimetric | |
96 wells | |
SCGB1A1 (Human) ELISA Kit | |
1 Kit | |
Plasma (Citrate, EDTA, Heparin), Serum | |
46pg/mL | |
96 Assays |