missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TNNI3 (Monkey) ELISA Kit
Brand: Abnova™ KA2445.1Kit
Additional Details : Weight : 0.02000kg
Description
TNNI3 (Monkey) ELISA Kit is an enzyme immunoassay for the quantitative determination of TNNI3 in monkey serum
Quantification
Specifications
TNNI3 (Monkey) ELISA Kit is for the quantitative measurement of TNNI3 in serum. | |
Quantification | |
TNNI3 (Monkey) ELISA Kit | |
Store Lyophilized Standard at -20°C. Store all other components at 4°C. |