Learn More
Abnova™ Varicella zoster virus IgG ELISA Kit
Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against Varicella zoster Virus
Brand: Abnova™ KA2262.1Kit
Additional Details : Weight : 0.02000kg
Specifications
Varicella zoster virus IgG ELISA Kit is intended for the detection of IgG antibody to VZV in human serum or plasma. | |
Qualitative | |
1 Kit | |
10μL |
Colorimetric | |
Varicella zoster virus IgG ELISA Kit | |
Plasma, Serum | |
Store the kit at 4°C. |