Learn More
Abnova™ Varicella zoster virus IgG ELISA Kit
Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against Varicella zoster Virus
Brand:  Abnova™ KA2262.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16057175
  • 2709.00 SEK / 1 set
Estimated Shipment
10-02-2021
Specifications
Varicella zoster virus IgG ELISA Kit is intended for the detection of IgG antibody to VZV in human serum or plasma.
Qualitative
1 Kit
10μL
Colorimetric
Varicella zoster virus IgG ELISA Kit
Plasma, Serum
Store the kit at 4°C.