missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Varicella zoster virus IgG ELISA Kit
Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against Varicella zoster Virus
POA
Specifications
Detection Method | Colorimetric |
---|---|
Product Type | Varicella zoster virus IgG ELISA Kit |
Content And Storage | Store the kit at 4°C |
Includes:
- 8-well break-away strips coated with specific antigen, 96 (8 x 12) wells
- Standard A 60 mIU (international (mili-) units), r.t.u., Ready to use, 1.3mL
- Standard B 200 mIU, Ready to use, 1.3mL
- Standard C 500 mIU, Ready to use, 1.3mL
- Standard D 1500 mIU, Ready to use, 1.3mL
- Standard E 10000 mIU, Ready to use, 1.3mL
- Anti-human IgG antibodies labelled with horseradish peroxidase, Ready to use, (Px-conjugate), 15mL
- Wash buffer, 10x concentarted, 125mL
- Dilution buffer, Ready to use, 125mL
- Chromogenic substrate (TMB substrate), Ready to use, 15mL
- Stop solution, Ready to use, 15mL
- Sealable pouch for unused strips, 1 piece
Specifications
Colorimetric | |
Varicella zoster virus IgG ELISA Kit | |
Store the kit at 4°C |