Läs mer
Brannan™ ABS Plastic Wall Mounted Clock
Beskrivning
Large dial wall clock with clear display makes this clock ideal for use in a variety of locations including schools, workplaces and in the home.
- Includes battery.
- ABS case
- Accurate quartz movement
- Diameter: 270mm.
Specifikationer
Specifikationer
| Produkttyp | Wall Clock |
| Tidsintervall | 12 hr. |
| Noggrannhet | Quartz movement |
| Visa | Stor urtavla |
| Material | ABS plastic |
| Diameter (metrisk) Yttre | 270 mm |
| Inkluderar | AA Battery |