Läs mer
ABT-239, MedChemExpress
Produktkod. 16495008
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16495008 10 mg -
16491247 50 mg -
16477737 5 mg 5mg
16423286 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16495008 Leverantör Medchem Express Leverantörsnummer HY12195
MedChemExpress ABT-239 is a novel, highly efficacious, non-imidazole class of H3R antagonist and a transient receptor potential vanilloid type 1 (TRPV1) antagonist.
ABT-239 is a novel, highly efficacious, non-imidazole class of H3R antagonist and a transient receptor potential vanilloid type 1 (TRPV1) antagonist.
Specifikationer
Kemiskt namn eller material ABT-239
Fysisk form Solid
CAS 460746-46-7
Färg Earth Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C22H22N2O
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : ≥ 100 mg/mL (302.65 mM) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER C[C@H]1N(CCC2=CC3=C(C=CC(C4=CC=C(C#N)C=C4)=C3)O2)CCC1
Molekylvikt (g/mol) 330.42
Formel vikt 330.42
Procent renhet 98.49%