Läs mer
Tocris Bioscience™ AC 265347
CaSR biased allosteric modulator
Brand: Tocris Bioscience™ 6165/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AC 265347 is a CaSR biased allosteric modulator (pEC50 = 7.8-8.1). Calcimimetic. Displays ability to bias signalling towards the accumulation of pERK1/2 and IP1. Reduces parathyroid hormone (PTH) levels in rat serum without inducing the release of calcitonin. Exhibits no significant activation of human GABAB or type I PTH receptors. Orally active.Specifikationer
| α-(2,4-Dimethylphenyl)-α-methyl-2-benzothiazolemethanol | |
| 1253901-26-6 | |
| >99% | |
| 10 mg |