missing translation for 'onlineSavingsMsg'
Läs mer

AC-ALA-PNA, TRC

Produktkod. 30240584
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30240584 500 mg -
30240838 250 mg -
30240995 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240584 Supplier Toronto Research Chemicals Supplier No. TRCA192438500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35978-75-7
Molekylformel C11H13N3O4
Molekylvikt (g/mol) 251.24
Synonym Acetylalanine 4-Nitroanilide
IUPAC-namn (2S)-2-acetamido-N-(4-nitrophenyl)propanamide
LEDER C[C@H](NC(=O)C)C(=O)Nc1ccc(cc1)[N+](=O)[O-]

Specifications

Kemiskt namn eller material AC-ALA-PNA
InChI formel InChI=1S/C11H13N3O4/c1-7(12-8(2)15)11(16)13-9-3-5-10(6-4-9)14(17)18/h3-7H,1-2H3,(H,12,15)(H,13,16)/t7-/m0/s1
Kvantitet 500 mg
Formel vikt 251.0906
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.