Läs mer
AC-ALA-PNA, TRC
Produktkod. 30240995
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30240995 2.5 g 2.5g
30240584 500 mg -
30240838 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1924382.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35978-75-7
Molekylformel C11H13N3O4
Molekylvikt (g/mol) 251.24
Synonym Acetylalanine 4-Nitroanilide
IUPAC-namn (2S)-2-acetamido-N-(4-nitrophenyl)propanamide
LEDER C[C@H](NC(=O)C)C(=O)Nc1ccc(cc1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material AC-ALA-PNA
InChI formel InChI=1S/C11H13N3O4/c1-7(12-8(2)15)11(16)13-9-3-5-10(6-4-9)14(17)18/h3-7H,1-2H3,(H,12,15)(H,13,16)/t7-/m0/s1
Kvantitet 2.5 g
Formel vikt 251.0906