missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ac-LEU-LEU-TYR-7-Amino-4-Trifluoromethylcoumarin
A fluorogenic substrate for Calpain
4298.30 SEK - 7610.80 SEK
Specifications
Color | White |
---|---|
Form | Solid |
For Use With (Application) | Cell Migration Analysis, Cell Signaling Analysis |
Molecular Weight (g/mol) | 660 |
Storage Requirements | Keep cool and dry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439875
|
MP Biomedicals™
03AFC11610 |
10mg |
4298.30 SEK
10mg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
11449875
|
MP Biomedicals™
03AFC11625 |
25mg |
7610.80 SEK
25mg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The AFC substrate is both chromogenic and fluorogenic. The specificity and sensitivity of AFC allows for the detection of enzyme activities in cell lysate and electrophoretic gels as the emission spectra of free and the conjugated AFC do not overlap. Excitation 400nm; Emission 505nm
Specifications
White | |
Cell Migration Analysis, Cell Signaling Analysis | |
Keep cool and dry |
Solid | |
660 |