missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ACADL (Human) Recombinant Protein
Human ACADL full-length ORF ( AAH39063, 1 a.a. - 430 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000033-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAARLLRGSLRVLGGHRAPRQLPAARCSHSGGEERLETPSAKKLTDIGIRRIFSPEHDIFRKSVRKFFQEEVIPHHSEWEKAGEVSREVWEKAGKQGLLGVNIAEHLGGIGGDLYSAAIVWEEQAYSNCSGPGFSIHSGIVMSYITNHGSEEQIKHFIPQMTAGKCIGAIAMTEPGAGSDLQGIKTNAKKDGSDWILNGSKVFISNGSLSDVVIVVAVTNHEAPSPAHGISLFLVENGMKGFIKGRKLHKMGLKAQDTAELFFEDIRLPASALLGEENKGFYYIMKELPQERLLIADVAISASEFMFEETRNYVKQRKAFGKTVAHLQTVQHKLAELKTHICVTRAFVDNCLQLHEAKRLDSATACMAKYWASELQNSVAYDCVQLHGGWGYMWEYPIAKAYVDARVQPIYGGTNEIMKELIAREIVFDK
Specifications
AAH39063 | |
acyl-Coenzyme A dehydrogenase, long chain | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACADL | |
GST |
33 | |
Wheat germ expression system | |
10 μg | |
ACAD4, FLJ94052, LCAD | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |