missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ACADL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582450
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: GEERLETPSA KKLTDIGIRR IFSPEHDIFR KSVRKFFQEE VIPHHSEWEK AGEVSREVWE KAGKQGLLGV NIAEHLGGIG GDLYSAAIVW EEQAYSNCSG PGFSIHSGIV MSYITNHGSE EQIKHFIPQM T Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 82%.
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia.
Specifikationer
| ACADL | |
| Polyclonal | |
| Unconjugated | |
| ACADL | |
| AA960361; ACAD4; ACADL; acetyl-Coenzyme A dehydrogenase, long-chain; ACOADA; Acyl Coenzyme A dehydrogenase, long chain; acyl-CoA dehydrogenase long chain; acyl-CoA dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long-chain; AU018452; C79855; LCAD; LCAD long chain acyl-CoA dehydrogenase; LOC100052267; long-chain acyl-CoA dehydrogenase; long-chain specific acyl-CoA dehydrogenase, mitochondrial; uncharacterized protein LOC100052267; zgc:55656 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11363, 25287, 33 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P15650, P28330, P51174 | |
| ACADL | |
| Recombinant protein corresponding to Human ACADL. Recombinant protein control fragment (Product #RP-88960). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering