Läs mer
Acamylophenine Dihydrochloride, TRC
Produktkod. 30316808
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30316808 500 mg
1 options
Produktkod. 30316808 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA589240500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acamylophenine Dihydrochloride
CAS 5892-41-1
Anteckningar om renhetsgrad HPLC
Molekylformel C19H32N2O2 . 2 (HCl)
Kvantitet 500 mg
Synonym 3-Methylbutyl [[2-(diethylamino)ethyl]amino](phenyl)acetate Dihydrochloride, Anafortin, Camylofin Dihydrochloride, Camylofin Hydrochloride, Camylofine Dihydrochloride, Camylofine Hydrochloride, Isopentyl alpha-[[2-(Diethylamino)ethyl]amino]phenylacetate Dihydrochloride
InChI formel InChI=1S/C19H32N2O2.2ClH/c1-5-21(6-2)14-13-20-18(17-10-8-7-9-11-17)19(22)23-15-12-16(3)4;;/h7-11,16,18,20H,5-6,12-15H2,1-4H3;2*1H
LEDER Cl.Cl.CCN(CC)CCNC(C(=O)OCCC(C)C)c1ccccc1
IUPAC-namn 3-methylbutyl 2-[2-(diethylamino)ethylamino]-2-phenylacetate;dihydrochloride
Molekylvikt (g/mol) 393.4
Formel vikt 392.1997