missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ACBD4 |
---|---|
Concentration | 0.62 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850525
|
Proteintech
20941-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840525
|
Proteintech
20941-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.Specifications
ACBD4 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q8NC06 | |
79777 | |
ACBD4 Fusion Protein Ag15045 | |
Primary | |
-20°C | |
ACBD4 |
0.62 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACBD4, HMFT0700 | |
ACBD4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |