Learn More
ACC1 Mouse anti-Human, Unconjugated, Clone: 1A11G10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67373-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839965
  • 4721.67 SEK / 150µL
Estimated Shipment: 19-04-2024
to see stock.
Description
Description
The ACC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACC1. This antibody recognizes human, mouse, rat antigen. The ACC1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ACC1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1A11G10
Unconjugated
150 μL
31
IgG2a