Learn More
ACC1 Mouse anti-Human, Unconjugated, Clone: 1A11G10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67373-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The ACC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACC1. This antibody recognizes human, mouse, rat antigen. The ACC1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ACC1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1A11G10 | |
Unconjugated | |
150 μL | |
31 | |
IgG2a |