Learn More
Thermo Scientific™ 1000mL BIOS Swinging Bucket Rotors
Efficiently and reliably perform your bioprocessing methods with the Thermo Scientific™ 6 x 1000mL or 8 x 1000mL swinging bucket rotors.
Brand: Thermo Scientific™ 12995
Additional Details : Weight : 1.00000kg
Description
The Thermo Scientific™ 1000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. These rotors, which are provided complete with buckets and bottles, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:- 8 x 1000mL (8L) capacity at 5,400 max rpm (8,500 x g max RCF) operation;
- 6 x 1000mL (6L) capacity at 4,600 max rpm (7,117 x g max RCF) operation
- Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 64% when compared with non-windshielded rotors
- Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
- 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
Seal Kit | |
1000mL Swinging Bucket Rotor Sets | |
Seal Kit for Cover for Round Bucket (sets of 6) |