Learn More
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Product Code. 15356617 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
3 Prong Clamp (60 mm Grip)
C-5, Clamp Holder
Heating Bath
Support Rod
Temperature Probe PT100
Temperature Probe PT100, SN-8-4
Transparent Shield (PC)
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15356617 3 Prong Clamp (60 mm Grip) Each
15386607 Support Rod Each
15376607 Transparent Shield (PC) Each
15366607 Heating Bath Each
15336617 Temperature Probe PT100, SN-8-4 Each
15346617 C-5, Clamp Holder Each
15366617 Temperature Probe PT100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15356617
  • ONLINE EXCLUSIVE PRICE
    213.00 SEK
    191.79 SEK / Paket med 1
    Save 21.21 SEK
    9% Av
Beräknad leverans: 21-04-2027
Optimize performance and flexibility
Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers. Optimize performance and flexibility
  • Accessory for use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Type 3 Prong Clamp (60 mm Grip)