Learn More
Thermo Scientific™ Accessories for the LP Vortex Mixer
Product Code. 15288874 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Rubber Platform
Tube Holder 12 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15288874 Tube Holder 12 mm Each
15248874 Rubber Platform Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15288874 Supplier Thermo Scientific™ Supplier No. 88880121
  • ONLINE EXCLUSIVE PRICE
    200.35 SEK
    186.12 SEK / Paket med 1
    Save 14.23 SEK
    7% Av
 Restricted Item
I lager
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
  •  Warranty: 2 years
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Holder 12 mm
Diameter (Metric) 12 mm
Description Tube Holder with Insert Retainer (dia. 12mm)
For Use With (Equipment) Vortex Mixer
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.