Learn More
Merck Accessories for YT31042HW Hazardous Waste Filtration System, 142 mm
Accessories for filter holders for sample preparation.
14575.00 SEK - 80760.00 SEK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Sterility | Non-sterile |
Type | Environmental Test Kits or Supplies |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10334951
|
Merck
YT3014257 |
Underdrain Support |
14575.00 SEK
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
12444277
|
Merck
YT310RAHW |
Rotary Agitator |
80760.00 SEK
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
Description
Accessories for filter holders for sample preparation.
YT3014257 Underdrain Support, PTFE-coatedYT310RAHW Rotary AgitatorSpecifications
Not Autoclavable | |
Non-sterile | |
Environmental Test Kits or Supplies |