Learn More
Merck Accessories for YY3009000 Filter Holder, 90 mm
Accessories for filter holders for sample preparation.
Brand: Merck YY3009054
4640.00 SEK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Accessories for filter holders for sample preparation.
AccessoriesXX4204708 Vent/Relief ValveYY3009053 O-rings, 90 mm filter housing, PTFEYY3009054 Support Screen, PTFE-coatedYY2209058 Underdrain Support, 90 mmYY2209059 Support LegsSpecifications
Not Autoclavable | |
90 mm filter holders | |
Non-sterile |
Support Screen | |
PTFE Coated 316 Stainless Steel | |
Support Screen |