Learn More
Thermo Scientific™ Acclaim™ 5μm PepMap™ μ-Precolumns™ Cartridge Columns
Very short microcolumns consisting of a cartridge holder and a set of disposable cartridges.
Brand: Thermo Scientific™ 160450
This item is not returnable.
View return policy
Description
Thermo Scientific™ Acclaim 5μm PepMap™ μ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a cartridge holder and a set of disposable cartridges.
- Used for sample pre-concentration, sample cleanup, or in various column-switching techniques
Specifications
Pre-Column Traps | |
LC/MS, ESI-MS, MALDI-MS | |
5 μm | |
100 Å | |
L1 | |
nanoLC | |
15% | |
Reversed Phase |
0.5 mm | |
15 mm | |
2 to 8 | |
450 m²/g | |
C18 | |
Silica | |
Peptide Analysis | |
Stainless Steel |