Learn More
Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns
Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a set of disposable cartridges. Cartridge holder sold separately (p/n 164650).
4442.10 SEK - 4693.02 SEK
Specifications
Column Format | Pre-Column Traps |
---|---|
Diameter (Metric) | 1 mm |
For Use With | LC/MS, ESI-MS, MALDI-MS |
Particle Size | 5 μm |
pH | 2 to 8 |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11362223
|
Thermo Scientific™
163592 |
5 μm | 1 mm | 5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11372223
|
Thermo Scientific™
163593 |
5 μm | 1 mm | 15 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a set of disposable cartridges. Cartridge holder sold separately (p/n 164650).
- For sample pre-concentration, sample cleanup, or in various column-switching techniques
- 300Å pore size
Specifications
Pre-Column Traps | |
LC/MS, ESI-MS, MALDI-MS | |
2 to 8 | |
100 m²/g | |
C18 | |
Silica | |
Peptide Analysis | |
Stainless Steel |
1 mm | |
5 μm | |
300 Å | |
L1 | |
nanoLC | |
9% | |
Reversed Phase |