missing translation for 'onlineSavingsMsg'
Läs mer

Thermo Scientific™ PepMap™ 300 C18 HPLC Columns

Produktkod. 13471208 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Längd (metrisk):
5 mm
150 mm
Diameter (metrisk):
0.075 mm
0.3 mm
1 mm
Kolumnformat:
Analytical Column
Analytical Column, NanoViper
Trap Cartridge
Förpackningsstorlek:
Paket med 5
Varje
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Längd (metrisk) Diameter (metrisk) Kolumnformat unitSize
13471208 150 mm 0.3 mm Analytical Column, NanoViper Varje
11312213 150 mm 0.075 mm Analytical Column Varje
13481208 150 mm 1 mm Analytical Column, NanoViper Varje
11342223 5 mm 0.3 mm Trap Cartridge Paket med 5
17224444 150 mm 0.075 mm Analytical Column, NanoViper -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 13471208 Leverantör Thermo Scientific™ Leverantörsnummer 164718

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Use this nanocolumn, packed for LC-MS/MS peptide mapping, protein identification, biomarker discovery, and systems biology applications.

This is the standard column for high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification. The Thermo Scientific™ Acclaim™ PepMap™ 300 C18 LC Columns is ideal for:

  • High-resolution peptide mapping of tryptic, natural, and synthetic peptides
  • LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology

High loading capacity

 

Acclaim PepMap columns are exceptionally suitable for the analysis of low abundant peptides often found in complex proteomics samples.

The Acclaim PepMap C18 300Å column 5μm Particle Size is available in 150mm lengths in 75, 300, 1000, and classic 75μm I.D. to allow for tuning throughput, sensitivity, and amount of sample available.

 

  • Superior protein and peptide separations
  • Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
  • Outstanding separation efficiency
  • Superior resolution and recovery
  • Easy installation with nanoViper™ Fingertight Fittings

Columns are available with Thermo Scientific™ Dionex™ nanoViper Fingertight Fittings:

  • Requires no tools
  • Provides virtually zero-dead-volume connections
  • Withstands pressures up to a 1000 bar
  • About Nano, capillary, and micro Acclaim PepMap columns

Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and micro columns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and micro column in a variety of chemistries for peptides and protein analysis.

TRUSTED_SUSTAINABILITY

Specifikationer

Kolumnformat Analytical Column, NanoViper
Diameter (metrisk) 0.3 mm
Längd (metrisk) 150 mm
Max. Tryck 7250 psi (500 bar)
Partikelstorlek 5 μm
pH 2 to 8
Porstorlek 300 Å
Ytarea 100 m²/g
Temperatur 60°C
USP typ L1
Stationär fas C18
Produktlinje nanoLC
Förpackningsmaterial Spherical, Fully Porous, Ultrapure Silica
Kolbelastning 9%
Kolumntyp Reversed Phase
Endcapped Yes
Fas Reversed Phase
Typ Analytical Column, NanoViper
Visa mer Visa mindre

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.