Läs mer
Aceclidine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Aceclidine Hydrochloride |
| CAS | 6109-70-2 |
| Molekylformel | C9 H15 N O2 . HCl |
| Kvantitet | 100 mg |
| Synonym | 1-Azabicyclo[2.2.2]octan-3-ol, 3-acetate, hydrochloride (1:1), 1-Azabicyclo[2.2.2]octan-3-ol, acetate (ester), hydrochloride (9CI), 3-Quinuclidinol, acetate (ester), hydrochloride (8CI), 3-Quinuclidinol, acetate, hydrochloride (7CI), (±)-3-Acetoxyquinuclidine hydrochloride, 3-Acetoxyquinuclidine hydrochloride, Aceclidine hydrochloride, Glaucostat |
| InChI formel | InChI=1S/C9H15NO2.ClH/c1-7(11)12-9-6-10-4-2-8(9)3-5-10;/h8-9H,2-6H2,1H3;1H |
| LEDER | Cl.O=C(OC1CN2CCC1CC2)C |
| IUPAC-namn | 1-azabicyclo[2.2.2]octan-3-yl acetate;hydrochloride |
| Molekylvikt (g/mol) | 205.68 |
| Formel vikt | 205.087 |