Läs mer
Acenocoumarol, TRC
Produktkod. 18052550
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18052550 10 mg
17939969 50 mg
18082150 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18052550 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA13080010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 152-72-7
Molekylformel C19 H15 N O6
Molekylvikt (g/mol) 353.33
Synonym 2H-1-Benzopyran-2-one, 4-hydroxy-3-[1-(4-nitrophenyl)-3-oxobutyl]-, Acenocoumarol (6CI), Coumarin, 3-(α-acetonyl-p-nitrobenzyl)-4-hydroxy- (8CI), (±)-Acenocoumarin, (±)-Acenocoumarol, (±)-Nicoumalone, (±)-p-Nitrowarfarin, 3-(Alpha-acetonyl-4-nitrobenzyl)-4-hydroxycoumarin, 3-(α-Acetonyl-4-nitrobenzyl)-4-hydroxycoumarin, 3-(α-Acetonyl-p-nitrobenzyl)-4-hydroxycoumarin, 3-(α-p-Nitrophenyl-β-acetylethyl)-4-hydroxycoumarin, 3-[2-Acetyl-1-(p-nitrophenyl)ethyl]-4-hydroxycoumarin, 3-[α-(4'-Nitrophenyl)-β-acetylethyl]-4-hydroxycoumarin, 3-[α-(p-Nitrophenol)-β-acetylethyl]-4-hydroxycoumarin, 4-Hydroxy-2-oxo-3-[3-oxo-1-(4-nitrophenyl)butyl]-2H-chromene, Acebron, Acenocoumarin, Acenokumarol, Acitrom, Ascumar, DL-3-(α-Acetonyl-4-nitrobenzyl)-4-hydroxycoumarin, G 23,350, G 23350, Minisintrom, Neo-Sintrom, Nicoumalone, Nitrowarfarin, Sincoumar, Sinkumar, Sinthrom, Sinthrome, Sintrom, Sintrom Mitis, Sintroma, Syncoumar, Syncumar, Syntrom, Trombostop, Zotil
IUPAC-namn 4-hydroxy-3-[1-(4-nitrophenyl)-3-oxobutyl]chromen-2-one
LEDER CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3ccc(cc3)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Acenocoumarol
Kvantitet 10 mg
InChI formel InChI=1S/C19H15NO6/c1-11(21)10-15(12-6-8-13(9-7-12)20(24)25)17-18(22)14-4-2-3-5-16(14)26-19(17)23/h2-9,15,22H,10H2,1H3
Formel vikt 353.0899