missing translation for 'onlineSavingsMsg'
Läs mer

Acetaldehyde-d4, TRC

Product Code. 30324944
Change view
Click to view available options
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30324944 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324944 Supplier Toronto Research Chemicals Supplier No. TRCA1326025G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Acetaldehyde-d4
CAS 1632-89-9
Molekylformel C2 2H4 O
Kvantitet 5 g
Synonym Acetaldehyde-1,2,2,2-d4 (ACI), Acetaldehyde-d4 (6CI, 7CI, 8CI, 9CI), 1,2,2,2-Tetradeuterioethanone, Ethanal-d4, Perdeuterioacetaldehyde, Perdeuteroacetaldehyde, Tetradeuteroacetaldehyde, Acetaldehyde-D4
InChI formel InChI=1S/C2H4O/c1-2-3/h2H,1H3/i1D3,2D
LEDER [2H]C(=O)C([2H])([2H])[2H]
IUPAC-namn 1,2,2,2-tetradeuterioethanone
Molekylvikt (g/mol) 48.08
Formel vikt 48.0513
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.