Learn More
Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog Number p-3756519
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
124-42-5
C2H6N2·ClH
94.54
MFCD00013016
WCQOBLXWLRDEQA-UHFFFAOYSA-N
acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
67170
ethanimidamide;hydrochloride
CC(=N)N.Cl
124-42-5
C2H6N2·ClH
94.54
MFCD00013016
WCQOBLXWLRDEQA-UHFFFAOYSA-N
acetamidine hydrochloride, acetimidamide hydrochloride, ethanimidamide hydrochloride, ethanimidamide, monohydrochloride, acetamidine hcl, ethanamidine hydrochloride, acetamidinium chloride, acediamine hydrochloride, ethenylamidine hydrochloride, acetamidine monohydrochloride
67170
ethanimidamide;hydrochloride
CC(=N)N.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 160°C to 170°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3C(=NH)NH2·HCl
Quantity 10 g
Beilstein 02, 185
Merck Index 15, 45
Solubility Information Solubility in water: 1g/mL. Other solubilities: soluble in alcohol,insoluble in acetone
Formula Weight 94.54
Percent Purity 98+%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Acetamidine hydrochloride