Läs mer
Acetaminophen-13C2, 15N1, TRC
Produktkod. 30133208
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133208 10 mg
17933245 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133208 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA16122810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 360769-21-7
Molekylformel C613C2H915NO2
Molekylvikt (g/mol) 154.14
Synonym Acetamide-13C2-15N, N-(4-hydroxyphenyl)- (9CI), Paracetamol-13C2-15N (acetamido-13C2-15N), Paracetamol-13C2,15N, N-(4-Hydroxyphenyl)acetamide-13C2-15N
IUPAC-namn N-(4-hydroxyphenyl)acetamide
LEDER [13CH3][13C](=O)[15NH]c1ccc(O)cc1
Specifikationer
Kemiskt namn eller material Acetaminophen-13C2, 15N1
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i1+1,6+1,9+1
Formel vikt 154.0671
Procent renhet >95%