Läs mer
Acetaminophen-cysteine, TRC
Produktkod. 30137797
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137797 100 mg
30009348 50 mg
30009668 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137797 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA161255100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64014-06-8
Molekylformel C11H14N2O3S
Molekylvikt (g/mol) 254.31
Synonym S-[4-(Acetylamino)phenyl]-L-cysteine;
IUPAC-namn (2R)-3-(4-acetamidophenyl)sulfanyl-2-aminopropanoic acid
LEDER O=C(O)[C@@H](N)CSC1=CC=C(NC(C)=O)C=C1
Specifikationer
Kemiskt namn eller material Acetaminophen-cysteine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C11H14N2O3S/c1-7(14)13-8-2-4-9(5-3-8)17-6-10(12)11(15)16/h2-5,10H,6,12H2,1H3,(H,13,14)(H,15,16)/t10-/m0/s1
Kvantitet 100 mg
Formel vikt 254.0725
Procent renhet >95%