Läs mer
Acetic Aceclofenac, TRC
Produktkod. 30322665
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30322665 5 mg
1 options
Produktkod. 30322665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1615505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetic Aceclofenac
CAS 1215709-75-3
Molekylformel C18 H15 Cl2 N O6
Kvantitet 5 mg
Synonym [[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetic Acid, Aceclofenac Imp. G (EP), Acetic Aceclofenac, Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, 2-(carboxymethoxy)-2-oxoethyl ester (ACI), 2-(Carboxymethoxy)-2-oxoethyl 2-[(2,6-dichlorophenyl)amino]benzeneacetate (ACI)
InChI formel InChI=1S/C18H15Cl2NO6/c19-12-5-3-6-13(20)18(12)21-14-7-2-1-4-11(14)8-16(24)27-10-17(25)26-9-15(22)23/h1-7,21H,8-10H2,(H,22,23)
LEDER OC(=O)COC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC-namn 2-[2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetyl]oxyacetic acid
Molekylvikt (g/mol) 412.22
Formel vikt 411.0276