Läs mer
Acetic acid-1-13C, TRC
Produktkod. 30179264
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179264 50 mg
30179173 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179264 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA14503050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1563-79-7
Molekylformel C13CH4O2
Molekylvikt (g/mol) 61.04
Synonym Acetic-1-13C acid (6CI,7CI,8CI,9CI), Acetic acid-1-13C, Acetic acid 1-13C, [1-13C]Acetic acid
IUPAC-namn acetic acid
LEDER C[13C](=O)O
Specifikationer
Kemiskt namn eller material Acetic acid-1-13C
Kvantitet 50 mg
InChI formel InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i2+1
Formel vikt 61.0245