Läs mer
Acetic Acid 3-Buten-1-yl Ester, TRC
Produktkod. 30316565
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30316565 250 mg
1 options
Produktkod. 30316565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA158600250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetic Acid 3-Buten-1-yl Ester
CAS 1576-84-7
Molekylformel C6H10O2
Kvantitet 250 mg
Synonym 1-Acetate-3-buten-1-ol, 1-Acetoxy-3-butene, 2-Vinylethyl Acetate, 3-Butenyl Acetate, 4-Acetoxy-1-butene, NSC 157648, 3-Buten-1-ol Acetate
InChI formel InChI=1S/C6H10O2/c1-3-4-5-8-6(2)7/h3H,1,4-5H2,2H3
LEDER CC(=O)OCCC=C
IUPAC-namn but-3-enyl acetate
Molekylvikt (g/mol) 114.14
Formel vikt 114.0681