missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals

Catalog Number p-4510523
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
631-61-8
C2H7NO2
77.08
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
631-61-8
C2H7NO2
77.08
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 114.0°C
Color White
Flash Point 136°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Linear Formula CH3CO2NH4
Quantity 1 kg
Beilstein 02,96
Fieser 01,38; 07,11
Merck Index 15,492
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity 98%
Grade Pure
Physical Form Adhering Crystals
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'