Learn More
Acetic Anhydride, Supelco™
Product Code. 15846820
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15846820 25 L 25 L
15803820 1 L 1 L
15813820 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15846820 Supplier Supelco Supplier No. 1000429025
 Restricted Item
Beräknad leverans: 24-11-2026
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
for analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-24-7
Grade Research
Molecular Formula (CH3CO)2O
Molecular Weight (g/mol) 102.09
Specifications
Melting Point -73°C
Density 1.08 g/cm3 at 20°C
MDL Number MFCD00008705
Quantity 25 L
Synonym Acetic anhydride, Acetyl acetate, Acetyl oxide, Acetic anhydride