missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetamide, 97%, Thermo Scientific Chemicals

Catalog Number p-4258735
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5977-14-0
GCPWJFKTWGFEHH-UHFFFAOYSA-N
acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
80077
CHEBI:28515
3-oxobutanamide
CC(=O)CC(=O)N
This item is not returnable. View return policy
5977-14-0
GCPWJFKTWGFEHH-UHFFFAOYSA-N
acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
80077
CHEBI:28515
3-oxobutanamide
CC(=O)CC(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -53°C to 56°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C4H7NO2
Linear Formula CH3C(O)CH2C(O)NH2
Quantity 100 g
Solubility Information Solubility in water: miscible
Molecular Weight (g/mol) 101.11
Formula Weight 101.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'