Läs mer
Acetochlor ESA Sodium Salt, TRC
Produktkod. 30317155
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30317155 25 mg
1 options
Produktkod. 30317155 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA16250825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetochlor Ethane Sulfonic Acid Sodium Salt
CAS 947601-84-5
Molekylformel C14 H20 N O5 S . Na
Kvantitet 25 mg
Synonym Acetochlor Ethane Sulfonic Acid Sodium Salt, Ethanesulfonic acid, 2-[(ethoxymethyl)(2-ethyl-6-methylphenyl)amino]-2-oxo-, sodium salt (1:1)
InChI formel InChI=1S/C14H21NO5S.Na/c1-4-12-8-6-7-11(3)14(12)15(10-20-5-2)13(16)9-21(17,18)19;/h6-8H,4-5,9-10H2,1-3H3,(H,17,18,19);/q;+1/p-1
LEDER [Na+].CCOCN(C(=O)CS(=O)(=O)[O-])c1c(C)cccc1CC
IUPAC-namn sodium;2-[N-(ethoxymethyl)-2-ethyl-6-methylanilino]-2-oxoethanesulfonate
Molekylvikt (g/mol) 337.37
Formel vikt 337.096