missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Laboratory Purity, Solveco
Click to view available options
Quantity:
5 L
Packaging:
HDPE Bottle
Description
Acetone is a very good solvent that dissolves many plastics, fats, oils and organic compounds in general. It is a colorless, volatile liquid with a distinctive odor. Acetone is very flammable and the vapors can be irritating to the eyes.
- Molecular Formula: CH3COCH3
- CAS: 67-64-1
- Non-Volatile Matter: <0.001
Specifications
CAS | 67-64-1 |
Color | Colorless |
Acidity | 0.003% max (titratable) |
Density | 0.790–0.798g/mL at 20°C |
Boiling Point | 56°C |
Quantity | 5 L |
Packaging | HDPE Bottle |
Refractive Index | 1.358–1.360 at 20°C |
Molecular Formula | C3H6O |
MDL Number | MFCD00008765 |
Show More |
Safety and Handling
Product Identifier
- Aceton
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'