missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile (15N) (IP >90%), TRC

Product Code. 30179655
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30179655 10 mg
30179322 50 mg
30178918 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30179655 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA16415810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14149-39-4
Molekylformel C2 H3 15N
Molekylvikt (g/mol) 42.04
Synonym Acetonitrile-15N (8CI, 9CI), Acetonitrile(CH3C15N), [15N]Acetonitrile
IUPAC-namn acetonitrile
LEDER CC#[15N]

Specifications

Kemiskt namn eller material Acetonitrile (15N)
Kvantitet 10 mg
InChI formel InChI=1S/C2H3N/c1-2-3/h1H3/i3+1
Formel vikt 42.0236
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.