Läs mer
(Acetonitrile)[4-tert-butyl-2-(diisopropylphosphino)-1-methyl-1H-imidazole-κN,κP](η5-cyclopentadienyl)ruthenium(1+) hexafluorophosphate, TRC
Produktkod. 30311001
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30311001 10 mg
1 options
Produktkod. 30311001 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA16415510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Acetonitrile)[4-tert-butyl-2-(diisopropylphosphino)-1-methyl-1H-imidazole-kN,kP](eta5-cyclopentadienyl)ruthenium(1+) hexafluorophosphate
CAS 930601-66-4
Molekylformel C21H35N3PRu . PF6
Kvantitet 10 mg
Synonym (Acetonitrile)[4-tert-butyl-2-(diisopropylphosphino)-1-methyl-1H-imidazole-κN,κP](η5-cyclopentadienyl)ruthenium(1+) hexafluorophosphate
InChI formel InChI=1S/C15H27NP.C5H10.C2H5N.F5P.FH.Ru/c1-11(2)17(12(3)4)14-9-13(10-16(14)8)15(5,6)7;1-2-4-5-3-1;1-2-3;1-6(2,3,4)5;;/h10-12H,1-8H3;1-5H2;2-3H,1H3;;1H;
LEDER FP(F)(F)(F)F.C/[C]=N\[Ru+]1C2=C([P](C(C)C)1C(C)C)N(C)C=C2C(C)(C)C.[F-].c3cccc3
IUPAC-namn Acetonitrile(cyclopentadienyl)[2-(di-i-propylphosphino)-4-(t-butyl)-1-methyl-1H-imidazole]ruthenium(II) hexafluorophosphate
Molekylvikt (g/mol) 606.53
Formel vikt 605.12