missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d3, for NMR, 99.8 atom % D, Thermo Scientific Chemicals

Catalog Number p-7214484
Click to view available options
Quantity:
10 mL
25 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-26-0
44.07
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-26-0
44.07

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8440 g/mL
Flash Point 2°C
Refractive Index 1.3430 to 1.3450
Specific Gravity 0.844
Viscosity 0.39 mPa.s (20°C)
Quantity 10 mL
Melting Point -46.0°C
Boiling Point 79.0°C to 81.0°C
Molecular Formula C2D3N
Linear Formula CD3CN
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 44.07
Grade NMR
Chemical Name or Material Acetonitrile-d3
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'