Learn More
Acetyl(2-pyridinylmethyl)carbamic Acid 2-Methoxy-3-[[(octadecylamino)carbonyl]oxy]propyl Ester, TRC
Product Code. 30315913
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30315913 25 mg 25mg
1 options
Product Code. 30315913 Supplier Toronto Research Chemicals Supplier No. TRCA18796525MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetyl(2-pyridinylmethyl)carbamic Acid 2-Methoxy-3-(((octadecylamino)carbonyl)oxy)propyl Ester
CAS 100488-81-1
Molecular Formula C32H55N3O6
Quantity 25 mg
InChI Formula InChI=1S/C32H55N3O6/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-24-34-31(37)40-26-30(39-3)27-41-32(38)35(28(2)36)25-29-22-19-21-23-33-29/h19,21-23,30H,4-18,20,24-27H2,1-3H3,(H,34,37)
SMILES CCCCCCCCCCCCCCCCCCNC(=O)OCC(COC(=O)N(Cc1ccccn1)C(=O)C)OC
IUPAC Name [2-methoxy-3-(octadecylcarbamoyloxy)propyl] N-acetyl-N-(pyridin-2-ylmethyl)carbamate
Molecular Weight (g/mol) 577.8
Formula Weight 577.4091